Documentos Principais
GBL622503
Grace Bio-Labs CoverWell perfusion chambers
3 chambers, port diam. 1.5 mm, chamber W × L × thickness 3 mm × 32 mm × 0.6 mm
Sinônimo(s):
leak proof enclosure for slides, microscope stick on multiwell chamber, reusable press to seal multiwell chamber, water tight multiple well chamber
Selecione um tamanho
40 EA
R$ 1.574,00
R$ 1.574,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
40 EA
R$ 1.574,00
About This Item
Código UNSPSC:
41122600
NACRES:
NB.22
R$ 1.574,00
Check Cart for Availability
Materiais
clear polycarbonate backing
red silicon wells
esterilidade
non-sterile
Características
wells 3
embalagem
pack of 40 ea
fabricante/nome comercial
Grace Bio-Labs 622503
câmara L × C × espessura
3 mm × 32 mm × 0.6 mm
diâmetro da porta
1.5 mm
poços
3
Descrição geral
CoverWell perfusion chambers press-to-seal covers form water-tight, multiwell cell incubation or cytochemistry chambers when pressed to coverslips or microscope slides. Reagents can be quickly added and removed through dual access ports without disturbing or cross-contaminating specimens. Chambers are available in five chamber configurations allowing up to eight different specimens or analytical tests on one coverglass.
CoverWell perfusion chambers form removable, water-tight multiwell enclosures by pressing to coverglass or microscope slides. Chambers are made of medical-grade silicone bonded to clear, UV-transparent plastic, and are removable for permanent mounting. The thin polycarbonate cover displays excellent transparency. The perfusion chambers are ideal for live cell imaging on upright or inverted microscopes at high magnification.
Informações legais
CoverWell is a trademark of Grace Bio-Labs, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)