EPS44I040010
Eppendorf® Dedicated Platform
for flasks 2L LBR, Innova(R) S44i
About This Item
Características
for flasks 2L LBR, Innova(R) S44i
fabricante/nome comercial
S44I040010
Eppendorf®
Categorias relacionadas
Descrição geral
Compatible with single unit of Eppendorf Innova S44i. For double stacking the stacking kit EPS44I041005 required in addition
Dedicated ppendorf shelves to hold various volume size flasks securely while shaking in Innova S44i.
Informações legais
Eppendorf is a registered trademark of Eppendorf AG
geralmente comprado junto com este produto