EP6731010855
Eppendorf® CellXpert® C170i CO2 Incubator
Copper interior, water/humidity monitoring, inner door with 4 door segments, handle left side, AC/DC input 120 V, US 2-pin plug (US/JP/South America)
About This Item
Código UNSPSC:
41104405
embalagem
pack of 1 ea
fabricante/nome comercial
Eppendorf® 6731010855
Descrição geral
CellXpert® C170i, inner door with 4 door segments, handle left side, water and humidity monitoring package, copper package, 100 – 127 V/50 – 60 Hz (US/JP/South America)
Características e benefícios
- water and humidity monitoring package
- handle left side
- inner door with 4 door segments
- 180° HTD (standard with C170i)
- copper package: chamber, water tray and four shelves
- 100 – 127 V, 50 – 60 Hz
Informações legais
CellXpert is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Escolha uma das versões mais recentes:
Certificados de análise (COA)