Pular para o conteúdo
Merck
Todas as fotos(1)

Documentos Principais

AV46301

Sigma-Aldrich

Anti-MRPS12 antibody produced in rabbit

affinity isolated antibody

Sinônimo(s):

Anti-MPR-S12, Anti-MT-RPS12, Anti-Mitochondrial ribosomal protein S12, Anti-RPMS12, Anti-RPS12, Anti-RPSM12

Faça loginpara ver os preços organizacionais e de contrato

Selecione um tamanho

100 μL
R$ 3.345,00

R$ 3.345,00


Previsão de entrega em01 de junho de 2025



Selecione um tamanho

Alterar visualização
100 μL
R$ 3.345,00

About This Item

Código UNSPSC:
12352203
NACRES:
NA.41

R$ 3.345,00


Previsão de entrega em01 de junho de 2025


fonte biológica

rabbit

Nível de qualidade

conjugado

unconjugated

forma do anticorpo

affinity isolated antibody

tipo de produto de anticorpo

primary antibodies

clone

polyclonal

Formulário

buffered aqueous solution

peso molecular

12 kDa

reatividade de espécies

human

concentração

0.5 mg - 1 mg/mL

técnica(s)

western blot: suitable

nº de adesão NCBI

nº de adesão UniProt

Condições de expedição

wet ice

temperatura de armazenamento

−20°C

Informações sobre genes

human ... MRPS12(6183)

Descrição geral

Mitochondrial ribosomal protein S12 (MRPS12), which is similar to bacterial ribosomal protein S12, is a eucaryote nucleus-encoded mitoribosomal component of mitochondrial translational apparatus that is translated on mitochondrial ribosomes.

Especificidade

Anti-MRPS12 polyclonal antibody reacts with canine, human, rat, and bovine mitochondrial ribosomal protein S12 proteins.

Imunogênio

Synthetic peptide directed towards the N terminal region of human MRPS12

Aplicação

Anti-MRPS12 polyclonal antibody is used to tag mitochondrial ribosomal protein S12 for detection and quantitation by Western blotting and in plasma by immunohistochemical (IHC) techniques. It is used as a probe to determine the roles of mitochondrial ribosomal protein S12 in mitochondrial ribosome structure and protein synthesis.

Ações bioquímicas/fisiológicas

Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. MRPS12 is the 28S subunit protein that belongs to the ribosomal protein S12P family. The protein is a key component of the ribosomal small subunit and controls the decoding fidelity and susceptibility to aminoglycoside antibiotics.Mammalian mitochondrial ribosomal proteins are encoded by nuclear genes and help in protein synthesis within the mitochondrion. Mitochondrial ribosomes (mitoribosomes) consist of a small 28S subunit and a large 39S subunit. They have an estimated 75% protein to rRNA composition compared to prokaryotic ribosomes, where this ratio is reversed. Another difference between mammalian mitoribosomes and prokaryotic ribosomes is that the latter contain a 5S rRNA. Among different species, the proteins comprising the mitoribosome differ greatly in sequence, and sometimes in biochemical properties, which prevents easy recognition by sequence homology. This gene encodes a 28S subunit protein that belongs to the ribosomal protein S12P family. The encoded protein is a key component of the ribosomal small subunit and controls the decoding fidelity and susceptibility to aminoglycoside antibiotics. The gene for mitochondrial seryl-tRNA synthetase is located upstream and adjacent to this gene, and both genes are possible candidates for the autosomal dominant deafness gene (DFNA4). Splice variants that differ in the 5′ UTR have been found for this gene; all three variants encode the same protein.

Sequência

Synthetic peptide located within the following region: LVPRLWATCSMATLNQMHRLGPPKRPPRKLGPTEGRPQLKGVVLCTFTRK

forma física

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Exoneração de responsabilidade

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Não está encontrando o produto certo?  

Experimente o nosso Ferramenta de seleção de produtos.

Código de classe de armazenamento

10 - Combustible liquids

Classe de risco de água (WGK)

WGK 3

Ponto de fulgor (°F)

Not applicable

Ponto de fulgor (°C)

Not applicable


Escolha uma das versões mais recentes:

Certificados de análise (COA)

Lot/Batch Number

Não está vendo a versão correta?

Se precisar de uma versão específica, você pode procurar um certificado específico pelo número do lote ou da remessa.

Já possui este produto?

Encontre a documentação dos produtos que você adquiriu recentemente na biblioteca de documentos.

Visite a Biblioteca de Documentos

Sonia Emperador et al.
Frontiers in genetics, 5, 469-469 (2015-02-03)
Several homoplasmic pathologic mutations in mitochondrial DNA, such as those causing Leber hereditary optic neuropathy or non-syndromic hearing loss, show incomplete penetrance. Therefore, other elements must modify their pathogenicity. Discovery of these modifying factors is not an easy task because

Nossa equipe de cientistas tem experiência em todas as áreas de pesquisa, incluindo Life Sciences, ciência de materiais, síntese química, cromatografia, química analítica e muitas outras.

Entre em contato com a assistência técnica