Documentos Principais
Z743184
Julabo® bath covers
lift-up transparent cover to fit Pura 30
Selecione um tamanho
1 EA
R$ 5.040,00
R$ 5.040,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 5.040,00
About This Item
Código UNSPSC:
41103706
NACRES:
NB.77
R$ 5.040,00
Check Cart for Availability
embalagem
pack of 1 ea
fabricante/nome comercial
Julabo 9970584
Descrição geral
The hinged cover for use with the PURA 30 water bath reduces the evaporation of the bath fluid and saves energy. The rising steam is led back into the bath. This means that the bath fluid remains in the bath and refill cycles are extended. An additional benefit is that the cover prevents contamination of the samples in the bath.
Informações legais
Julabo is a registered trademark of Julabo USA, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)