Documentos Principais
Z741823
Extech Temperature, Humidity Datalogger
Kit with PC Interface
About This Item
Código UNSPSC:
41100000
NACRES:
NB.79
Características
CE compliant
resolution 0.1 %RH
resolution 0.1 °F/C
(WEEE Compliant)
embalagem
pkg of 1 ea
fabricante/nome comercial
Extech Instruments 42275
Parâmetros
-40-185 °F temperature (-40-85 °C)
tamanho
4.9 in. × 3.8 in. × 2 in., 123.8x95.3x51mm
peso
9 oz254
Descrição geral
Compact, powerful and easy-to-use dataloggers record up to 16,000 readings. Multiple Dataloggers can be programmed and data downloaded from one single docking station. USB cable and software included to easily download collected data to your PC for further analysis.
Escolha uma das versões mais recentes:
Certificados de análise (COA)