Documentos Principais
Z673471
LAB-AIRE® II non-electrical drying racks
wall mounted, single sided, 3 tier
Selecione um tamanho
1 EA
R$ 2.152,00
R$ 2.152,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 2.152,00
About This Item
Código UNSPSC:
41103200
NACRES:
NB.35
R$ 2.152,00
Check Cart for Availability
Características
no. of pegs 57
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art F18933-0013
C × L × A
14.75 in. × 23.4 in. × 5 in.
Descrição geral
Space in laboratories is always in high demand making the small footprint of the Scienceware® Lab-Aire II Drying Racks a practical solution for drying and storing labware. With a choice of benchtop or wall-mountable models in varying heights, you′re sure to find the rack that meets your specific needs.
All models come with three size and length rugged V-channel pegs that can be moved and repositioned to hold a variety of labware, including: beakers, flasks, bottles, test tubes and cylinders.
Additional accessory pegs, (19 per bag, small, medium or large) are available to reconfigure the rack for specific size and weight labware. Accommodates an assortment of glass and plastic labware by easily changing peg locations.
  • High storage capacity - up to 114 peg positions for all types of labware
  • Single-sided benchtop models fit against a wall, conserving bench space
  • Double-sided benchtop models provide twice the capacity with only a slightly larger footprint
  • Wall-mount models to save bench space
Informações legais
Lab-Aire is a registered trademark of SP Scienceware, a division of SP
Scienceware is a registered trademark of SP Scienceware, a division of SP
Escolha uma das versões mais recentes:
Certificados de análise (COA)