Documentos Principais
Z568759
Simriz® high-performance O-ring for QianCap
25-S
About This Item
Código UNSPSC:
41121815
NACRES:
NB.51
fabricante/nome comercial
Q Labtech
Descrição geral
Have excellent compatibility with most of the chemicals, including inorganic and organic acids and bases, alcohols, aliphatic and chlorinated hydrocarbons, aromatics, ketones, esters, ethers and aldehydes and fuel oils.
Informações legais
QianCap is a trademark of Q Labtech LLC
Simriz is a registered trademark of Carl Freudenburg KG
Escolha uma das versões mais recentes:
Certificados de análise (COA)