Z549312
Aldrich® anticlimb splash-guard adapter with fritted disc
female joint: ST/NS 29/32, male joint: ST/NS 14/20, capacity 250 mL
About This Item
Código UNSPSC:
41121815
Junta
female joint: ST/NS29/32
male joint: ST/NS14/20
capacidade
250 mL
tamanho de poro
70-100 μm porosity
Descrição geral
These traps greatly reduce carry-over from foaming or bumping. Because the venting hole is located at the top, the foam can be prevented from climbing higher into the condenser. The fritted disc prevents particulates from entering the condenser.
Informações legais
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Escolha uma das versões mais recentes:
Certificados de análise (COA)