Documentos Principais
Z202509
Sigma-Aldrich
Mini Diazald® apparatus
joint: ST/NS 19/26 Clear-Seal joints, for generating 1 to 50 mmol of diazomethane
Selecione um tamanho
1 EA
R$ 5.303,00
R$ 5.303,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 5.303,00
About This Item
Código UNSPSC:
41104812
NACRES:
NB.43
R$ 5.303,00
Check Cart for Availability
Junta
joint: ST/NS19/26 Clear-Seal joints
Categorias relacionadas
Descrição geral
For technical and safety information about Diazald and Aldrich diazomethane generators see Technical Information Bulletin AL-180.
The mini Diazald apparatus bridges the gap between the System 45 diazomethane generator Z411736 (for preparing <1 mmol of diazomethane) and the Diazald kit (for preparing ~100 mmol of diazomethane). It consists of a reaction vessel and a condenser in one compact unit. This apparatus features a cold-finger in place of a water-jacketed condenser. When filled with dry ice/acetone slush, the condenser efficiently prevents dangerous diazomethane/ether vapor from escaping. A separatory funnel, Z100382 and a round-bottom receiving flask, Z100358, (must be equipped with Clear-Seal joints) are the only other glassware needed and can be purchased separately.
NOTE: Sigma-Aldrich is unable to supply certain Diazomethane precursors in some countries. Check availability of starting materials prior to glassware purchase.
Informações legais
Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany
Escolha uma das versões mais recentes:
Certificados de análise (COA)