Documentos Principais
S853755
2-(2-CHLOROPHENOXY)ETHYL N-(4-METHYL-3-NITROPHENYL)CARBAMATE
AldrichCPR
Selecione um tamanho
50MG
R$ 61,00
R$ 61,00
Previsão de entrega em22 de maio de 2025
Selecione um tamanho
Alterar visualização
50MG
R$ 61,00
About This Item
Fórmula linear:
C16H15ClN2O5
Número CAS:
Peso molecular:
350.761
Número MDL:
R$ 61,00
Previsão de entrega em22 de maio de 2025
Outras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Escolha uma das versões mais recentes:
Certificados de análise (COA)