Documentos Principais
R521493
6,7-DIMETHYL-4-OXA-TRICYCLO(4.3.0.0(3,7))NONANE-3-CARBOXYLIC ACID PHENYLAMIDE
AldrichCPR
Selecione um tamanho
10MG
R$ 61,00
R$ 61,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
10MG
R$ 61,00
About This Item
Fórmula linear:
C17H21NO2
Número CAS:
Peso molecular:
271.362
Número MDL:
R$ 61,00
Check Cart for Availability
Outras notas
Please note that Sigma-Aldrich provides this product to early discovery researchers as part of a collection of rare and unique chemicals. Sigma-Aldrich does not collect analytical data for this product. Buyer assumes responsibility to confirm product identity and/or purity. All sales are final.
NOTWITHSTANDING ANY CONTRARY PROVISION CONTAINED IN SIGMA-ALDRICH'S STANDARD TERMS AND CONDITIONS OF SALE OR AN AGREEMENT BETWEEN SIGMA-ALDRICH AND BUYER, SIGMA-ALDRICH SELLS THIS PRODUCT AS-IS AND MAKES NO REPRESENTATION OR WARRANTY WHATSOEVER WITH RESPECT TO THIS PRODUCT, INCLUDING ANY (A) WARRANTY OF MERCHANTABILITY; (B) WARRANTY OF FITNESS FOR A PARTICULAR PURPOSE; OR (C) WARRANTY AGAINST INFRINGEMENT OF INTELLECTUAL PROPERTY RIGHTS OF A THIRD PARTY; WHETHER ARISING BY LAW, COURSE OF DEALING, COURSE OF PERFORMANCE, USAGE OF TRADE OR OTHERWISE.
Escolha uma das versões mais recentes:
Certificados de análise (COA)