Documentos Principais
PL800SHELFI
Plas-Labs transport support cart
sliding shelf package - single station
Selecione um tamanho
1 EA
R$ 18.530,00
R$ 18.530,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 18.530,00
About This Item
Código UNSPSC:
41123300
NACRES:
NB.35
R$ 18.530,00
Check Cart for Availability
A × L × P
18 in. × 18 in. × 10 in.
Descrição geral
Suspended sliding shelf for single operator units. Culture plate capacity is about 340. Fabricated of .375 in. thick clear acrylic, with shelves spaced at 7 in. apart. The entire assembly is suspended from the chamber top to keep the floor area uncluttered.
All carts have anti-scuff casters including brakes and locks. Carts also feature a cushioning system that minimizes vibration, shocks, and wear and tear on chambers.
Escolha uma das versões mais recentes:
Certificados de análise (COA)