Documentos Principais
DWK73802-33430
KIMBLE® Solid Cap
black phenolic cap (with rubber liner), cap for 33-400
Selecione um tamanho
1 EACH
R$ 2.986,00
R$ 2.986,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EACH
R$ 2.986,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.11
R$ 2.986,00
Check Cart for Availability
Materiais
PTFE/rubber liner
black phenolic cap (with rubber liner)
esterilidade
non-sterile
Características
class A
embalagem
case of 144 ea
fabricante/nome comercial
Kimble® 73802-33430
conector
cap for 33-400
Descrição geral
Class A Caps with PTFE-Faced Rubber Liners highly resistant to temperature and steam during autoclaving
  • Specially formulated phenolic cap material
  • Autoclavable
  • White rubber liners
  • Excellent for general laboratory use
Identified in ASTM Specification E982, Class A requirement
Características e benefícios
  • Specially formulated phenolic resin and liner adhesive to withstand the effects of repeated autoclaving
  • PTFE faced/general purpose white rubber liners are highly resistant to chemical effects
  • Excellent for general laboratory use
  • Identified in ASTM Specification E982, Class A requirements
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)