Documentos Principais
DWK218064455
DURAN® graduated amber laboratory bottle
with cap, round amber glass bottle, bottle capacity (500 mL), non-sterile
Sinônimo(s):
218064455, DURAN amber bottle, Laboratory bottle
Selecione um tamanho
10 EACH
R$ 5.897,00
R$ 5.897,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
10 EACH
R$ 5.897,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.17
R$ 5.897,00
Check Cart for Availability
Materiais
round amber glass bottle
esterilidade
non-sterile
Características
with cap
embalagem
case of 10 ea
fabricante/nome comercial
DURAN® 218064455
capacidade do frasco
(500 mL)
Descrição geral
Standardised GL-thread and matching cap system for practicularly tight sealing and simple, clean pouring out. The only external application of the colour ensures that internal surfaces have standard DURAN glass properties.
Very durable and chemically resistant amber colour. Easy-to-read scale with graduation marks to estimate the content. Fired-on, highly durable white ceramic print. Sizes 10 and 25 ml with specially shaped glass edge for improved pouring out whereby an additional plastic pouring ring is not required.
Informações legais
DURAN is a registered trademark of DWK Life Sciences
Escolha uma das versões mais recentes:
Certificados de análise (COA)