Documentos Principais
DWK14396-1000
KIMBLE® KIMAX® Media bottle
without cap, round transparent borosilicate glass bottle, bottle capacity (1,000 mL), non-sterile
Selecione um tamanho
10 EA
R$ 1.636,00
R$ 1.636,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
10 EA
R$ 1.636,00
About This Item
Código UNSPSC:
41121800
NACRES:
NB.43
R$ 1.636,00
Check Cart for Availability
Materiais
round transparent borosilicate glass bottle
esterilidade
non-sterile
Características
without cap
embalagem
case of 10 ea
fabricante/nome comercial
Kimble® 14396-1000
capacidade do frasco
(1,000 mL)
conector
GL 45 neck
GL 45
Descrição geral
GL 45 Media Bottles ideal for general laboratory use including mixing, storing or transporting culture media, chemicals or solvents.
  • Enhanced graduations and marking spot made with chemically resistant white enamel paint
  • 30 mm ID opening
  • Autoclavable
  • Supplied without caps or with linerless GL 45 screw thread caps
  • Replaceable clear (natural) drip-free polypropylene pour ring is included on each bottle
  • Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements
Informações legais
KIMBLE is a registered trademark of DWK Life Sciences
Kimax is a registered trademark of Gerresheimer Glass, Inc.
Escolha uma das versões mais recentes:
Certificados de análise (COA)