Documentos Principais
BAF132040001
Cover for glass disposal carton
autoclavable
Selecione um tamanho
1 EA
R$ 371,00
R$ 371,00
Check Cart for Availability
Selecione um tamanho
Alterar visualização
1 EA
R$ 371,00
About This Item
Código UNSPSC:
41100000
NACRES:
NB.35
R$ 371,00
Check Cart for Availability
Características
autoclavable
embalagem
pack of 1 ea
fabricante/nome comercial
Bel-Art F13204-0001
Descrição geral
Dishwasher-safe and autoclave-safe, the reusable Disposal Carton Cover opens quickly and easily by pressing the built-in release tab - no batteries required! The cover fits any disposal carton with a 30.4 x 30.4 cm (12 x 12 in.) opening.
  • Opens with a Simple Touch
  • Sanitary and reusable
  • Green center panel matches printing on glass disposal cartons providing fast visual recognition of the waste stream
  • Features smooth 10.1 x 10.1 cm (4 x 4 in.) area for applying labels
  • Provides a wider opening 20.3 x 24.1 cm (8 in. x 9 1/2 in.) permitting safer and more sanitary deposit of wastes into the container
Escolha uma das versões mais recentes:
Certificados de análise (COA)