Documentos Principais
BAF131741008
Cleanware self-adhesive waste bags
W × L 203 mm (8 in.) × 254 mm (10 in.)
Selecione um tamanho
50 EA
R$ 224,00
R$ 224,00
Previsão de entrega em16 de abril de 2025
Selecione um tamanho
Alterar visualização
50 EA
R$ 224,00
About This Item
Código UNSPSC:
24111500
NACRES:
NB.35
R$ 224,00
Previsão de entrega em16 de abril de 2025
embalagem
pack of 50 ea
fabricante/nome comercial
Bel-Art F13174-1008
L × C
203 mm (8 in.) × 254 mm (10 in.)
Descrição geral
These disposable waste bags feature an adhesive strip across the top so they can be hung from walls, cabinets, or virtually anywhere freeing bench or hood space. The adhesive backing strip is easily peeled off, and the bag can then be hung where needed. The back panel is longer than the front for easy opening. Maximum weight of contents should not exceed 1.36 kg (3 lb.). Packed 50 bags per pack
  • Convenient, hang-anywhere design
  • Simply peel off the backing and hang close to your work area
  • Made of High Molecular Weight High Density (HMHD) Polyethylene
  • Steam autoclavable at 121 ºC (250 ºF)
Informações legais
Escolha uma das versões mais recentes:
Certificados de análise (COA)