Key Documents
TMO4114-0038
Nalgene® vented sterile caps for Erlenmeyer flasks
high-density polyethylene, 125/250 mL flasks, pore size 0.2 μm, case of 12 ea
Synonym(s):
vented closure, vented single use closure
Select a Size
1 EACH
R$875.00
R$875.00
Check Cart for Availability
Select a Size
Change View
1 EACH
R$875.00
About This Item
UNSPSC Code:
41121804
NACRES:
NB.13
R$875.00
Check Cart for Availability
material
high-density polyethylene
sterility
sterile
feature
vented
packaging
case of 12 ea
manufacturer/tradename
Nalgene 4114-0038
pore size
0.2 μm pore size
General description
Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific Nalgene Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
Hold and sterilize refuse, clothing, equipment, labware and contaminated material with Thermo Scientific Nalgene Large Polypropylene Waste Containers with Cover. These containers feature a secure, friction-fit closure and molded-in handgrips. They meet OSHA Standard 29 CFR Part 1910.1030 for use as protection against bloodborne pathogens.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)