Skip to Content
Merck
All Photos(1)

Key Documents

CLS139545LTMC

Vented cap for Corning® media storage bottles

gray, GL 45

Synonym(s):

Cap for Corning Bottles, Corning Vented Cap, Vented Cap for Storage Bottles

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
R$390.00

R$390.00


Check Cart for Availability


Select a Size

Change View
10 EA
R$390.00

About This Item

UNSPSC Code:
41121800
NACRES:
NB.43

R$390.00


Check Cart for Availability

material

polypropylene

manufacturer/tradename

Corning 1395-45LTMC

color

gray

fitting

GL 45

Features and Benefits

  • Vented cap made up of polypropylene
  • Caps have a hydrophobic polytetrafluoroethylene (PTFE) membrane that allows contamination-free sterile gas exchange
  • To ensure efficient sealing, the cap has a GL45 thread and a linerless plug seal design
  • Autoclavable

Legal Information

Corning is a registered trademark of Corning, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service