Skip to Content
Merck
All Photos(1)

Key Documents

SCP0157

Sigma-Aldrich

Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP)

Synonym(s):

GRGDSP Peptide, RGD Peptide

Sign Into View Organizational & Contract Pricing

Select a Size

5 MG
R$2,109.00

R$2,109.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 MG
R$2,109.00

About This Item

Empirical Formula (Hill Notation):
C22H37N9O10
Molecular Weight:
587.58
UNSPSC Code:
12352209
NACRES:
NA.32

R$2,109.00


Check Cart for Availability

Request a Bulk Order

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥60%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Gly-Arg-Gly-Asp-Ser-Pro

General description

Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP) is a bioactive molecule/peptide derived from fibronectin.[1]

Application

Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP) has been used:
  • in coating well plate to monitor cell attachment by crystal violet staining[2]
  • to treat baby hamster kidney (BHK-21) cells to study the role of Arg-Gly-Asp (RGD) motif in the replication of Japanese encephalitis virus (JEV)[3]
  • to block RGD-dependent integrins or integrin signaling in INA-6 cells[4]
  • to study the role of the RGD-binding integrins in Gal-3-mediated cell adhesion[5]

Biochem/physiol Actions

Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP) can stimulate endothelialization by modifying the surface of cardiovascular implants. The major cell- binding domain of GRGDSP helps to promote cell adhesion on a wide range of substrates.[1]
Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP) is used as a soluble integrin-blocking RGD-based peptide. GRGDSP is used widely together with other RGD peptides in integrin research.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shyam Patel et al.
Journal of biomedical materials research. Part A, 83(2), 423-433 (2007-04-25)
Vascular endothelium plays an important role in preventing thrombogenesis. Bioactive molecules such as fibronectin-derived peptide Gly-Arg-Gly-Asp-Ser-Pro (GRGDSP) can be used to modify the surface of cardiovascular implants such as vascular grafts to promote endothelialization. Here we conjugated GRGDSP peptide to
Wenchun Fan et al.
Research in veterinary science, 111, 67-74 (2017-01-04)
Japanese encephalitis virus (JEV) is a mosquito-borne flavivirus that is one of the major causes of viral encephalitis diseases worldwide. The JEV envelope protein facilitates viral entry, and its domain III contains an Arg-Gly-Asp (RGD) motif, that may modulate JEV
Ines Lauria et al.
Acta biomaterialia, 62, 317-327 (2017-09-03)
Bionanoparticles based on filamentous phages or flexuous viruses are interesting candidates for meeting the challenges of tailoring biomineralization in hydrogel-based bone tissue substitutes. We hypothesized that hydroxyapatite crystal nucleation and matrix mineralization can be significantly increased by mineralization-inducing (MIP) and
Julia Dotterweich et al.
Cell communication and signaling : CCS, 12, 36-36 (2014-06-27)
CCN family member 1 (CCN1), also known as cysteine-rich angiogenic inducer 61 (CYR61), belongs to the extracellular matrix-associated CCN protein family. The diverse functions of these proteins include regulation of cell migration, adhesion, proliferation, differentiation and survival/apoptosis, induction of angiogenesis
Antonín Sedlář et al.
International journal of molecular sciences, 22(10) (2021-06-03)
Galectin-3 (Gal-3) is a β-galactoside-binding protein that influences various cell functions, including cell adhesion. We focused on the role of Gal-3 as an extracellular ligand mediating cell-matrix adhesion. We used human adipose tissue-derived stem cells and human umbilical vein endothelial

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service