Skip to Content
Merck
All Photos(1)

Key Documents

AFM-71855-15-10

Sigma-Aldrich

Muscovite Mica for AFM

(V-1 Quality), size 15 mm × 15 mm, pack of 10

Synonym(s):

Muscovite Mica for AFM

Sign Into View Organizational & Contract Pricing

Select a Size

10 PKG
R$225.00

R$225.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 PKG
R$225.00

About This Item

UNSPSC Code:
41111761
NACRES:
NA.23

R$225.00


Check Cart for Availability

Request a Bulk Order

form

solid

packaging

pack of 10

size

15 mm × 15 mm

thickness

0.15-0.2 mm

Looking for similar products? Visit Product Comparison Guide

General description

Mica, 15mmx15mm, thickness 0.15-0.2 mm, V-1 grade, Clear - Hard, of uniform color, nearly flat, free of all stains, foreign inclusion, cracks, and other similar defects. Perfect for AFM substrate

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

  1. Can this be used as a substrate for thin film deposition?

    1 answer
    1. Yes, this product is likely suitable for use as a substrate for thin film deposition. However, since it is primarily designed for Atomic Force Microscopy (AFM) and has not been specifically tested for thin film deposition processes, its performance will depend on the experimental condition of certain deposition technique. Further testing would be necessary to fully assess its suitability for thin film applications.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service