CLS354638
Corning® Vented Screw Cap
for use with Falcon® 75cm Flasks, sterile
Synonym(s):
Vented Screw Cap
About This Item
Recommended Products
material
PE flask
sterility
sterile
packaging
pack of 100 ea (10 packs of 10)
manufacturer/tradename
Corning 354638
compatibility
for use with Falcon® 75cm Flasks
Looking for similar products? Visit Product Comparison Guide
General description
Application
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service