Skip to Content
Merck
All Photos(1)

Key Documents

Y0000442

Didanosine impurity G

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2′,3′-Dideoxyadenosine, ddA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H13N5O2
CAS Number:
Molecular Weight:
235.24
Beilstein:
619924
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

didanosine

manufacturer/tradename

EDQM

mp

181-184 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Nc1ncnc2n(cnc12)[C@H]3CC[C@@H](CO)O3

InChI

1S/C10H13N5O2/c11-9-8-10(13-4-12-9)15(5-14-8)7-2-1-6(3-16)17-7/h4-7,16H,1-3H2,(H2,11,12,13)/t6-,7+/m0/s1

InChI key

WVXRAFOPTSTNLL-NKWVEPMBSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Didanosine impurity G EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anabolism and mechanism of action of Ro24-5098, an isomer of 2',3'-dideoxyadenosine (ddA) with anti-HIV activity.
K B Frank et al.
Annals of the New York Academy of Sciences, 616, 408-414 (1990-01-01)
S Rachel Skinner et al.
Lancet (London, England), 384(9961), 2213-2227 (2014-09-06)
Although adolescent girls are the main population for prophylactic human papillomavirus (HPV) vaccines, adult women who remain at risk of cervical cancer can also be vaccinated. We report data from the interim analysis of the ongoing VIVIANE study, the aim
Nicole M Warrington et al.
Cancer research, 75(1), 16-21 (2014-11-09)
Identifying modifiers of glioma risk in patients with type I neurofibromatosis (NF1) could help support personalized tumor surveillance, advance understanding of gliomagenesis, and potentially identify novel therapeutic targets. Here, we report genetic polymorphisms in the human adenylate cyclase gene adenylate
Amit Singhal et al.
Science translational medicine, 6(263), 263ra159-263ra159 (2014-11-21)
The global burden of tuberculosis (TB) morbidity and mortality remains immense. A potential new approach to TB therapy is to augment protective host immune responses. We report that the antidiabetic drug metformin (MET) reduces the intracellular growth of Mycobacterium tuberculosis

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service