Skip to Content
Merck
All Photos(1)

Key Documents

Y0000441

Didanosine impurity A,

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Hypoxanthine, 6-Hydroxypurine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H4N4O
CAS Number:
Molecular Weight:
136.11
Beilstein:
5811
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

pharmaceutical primary standard

API family

didanosine

manufacturer/tradename

EDQM

mp

>300 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

O=C1NC=Nc2nc[nH]c12

InChI

1S/C5H4N4O/c10-5-3-4(7-1-6-3)8-2-9-5/h1-2H,(H2,6,7,8,9,10)

InChI key

FDGQSTZJBFJUBT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Hypoxanthine is a nutrient additive for a variety of cell culture applications involving bacterial, parasite (Plasmodium falciparum) and animal cells. Hypoxanthine is a component of selection media used in hybridoma technologies.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypoxanthine as an indicator of hypoxia: its role in health and disease through free radical production.
O D Saugstad
Pediatric research, 23(2), 143-150 (1988-02-01)
Matthias M Heck et al.
European urology, 66(2), 222-229 (2013-03-08)
To determine the anatomic extent of pelvic lymph node dissection (PLND) in prostate cancer (PCa) patients at the time of radical prostatectomy (RP), knowledge about the topography of lymph node (LN) metastases is required. Because small-volume LN metastases may be
Chia-Tung Wu et al.
Cardiovascular research, 102(3), 487-496 (2014-03-07)
Fibroblasts, which play an important role in cardiac function/dysfunction, including arrhythmogenesis, have voltage-dependent (Kv) currents of unknown importance. Here, we assessed the differential expression of Kv currents between atrial and ventricular fibroblasts from control dogs and dogs with an atrial
Ian A Lewis et al.
PLoS genetics, 10(1), e1004085-e1004085 (2014-01-07)
Drug resistant strains of the malaria parasite, Plasmodium falciparum, have rendered chloroquine ineffective throughout much of the world. In parts of Africa and Asia, the coordinated shift from chloroquine to other drugs has resulted in the near disappearance of chloroquine-resistant
Katie May Slattery et al.
Medicine and science in sports and exercise, 46(6), 1114-1123 (2014-03-01)
This investigation examined the ergogenic effect of short-term oral N-acetylcysteine (NAC) supplementation and the associated changes in redox balance and inflammation during intense training. A double-blind randomized placebo-controlled crossover design was used to assess 9 d of oral NAC supplementation

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service