Skip to Content
Merck
All Photos(1)

Key Documents

70181

Millipore

Antibiotic Agar No 1

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Penassay Seed Agar, Grove and Randall Antibiotic Agar No. 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171504
NACRES:
NA.74

sterility

non-sterile

Quality Level

100
200

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

agar, 15.0 g/L
dextrose, 1.0 g/L
meat extract, 1.5 g/L
peptone (from casein), 4.0 g/L
peptone (from meat), 6.0 g/L
yeast extract, 3.0 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

final pH

6.5±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

suitability

Micrococcus spp.
nonselective for Bacillus spp.
nonselective for Bordetella spp.
nonselective for Klebsiella spp.
nonselective for Pseudomonas spp.
nonselective for Staphylococcus spp.
nonselective for bacteria (General Media)

Application

Used in antibiotic assay work (USP, FDA, Ph Eur); used as seed agar, e.g. with S. aureus for the assay of kanamycin or as base agar, e.g. for assaying chloramphenicol.

Preparation Note

Suspend 30.5 g in 1 litre of distilled water and bring to the boil to dissolve completely. Sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

D.C. Grove et al.
Assay Methods of Antibiotics, Med. Encycl. (1955)
Jasmine K Ahluwalia et al.
Retrovirology, 5, 117-117 (2008-12-24)
Cellular miRNAs play an important role in the regulation of gene expression in eukaryotes. Recently, miRNAs have also been shown to be able to target and inhibit viral gene expression. Computational predictions revealed earlier that the HIV-1 genome includes regions

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service