Skip to Content
Merck
All Photos(1)

Key Documents

70184

Millipore

Antibiotic Broth

suitable for microbiology, NutriSelect® Plus

Synonym(s):

Antibiotic Medium No. 3, Assay Broth, Penassay Broth, Grove and Randall Antibiotic Broth

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$1,166.00

R$1,166.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

Select a Size

Change View
500 G
R$1,166.00

About This Item

UNSPSC Code:
41171504
NACRES:
NA.74

R$1,166.00


Estimated to ship onMay 29, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

composition

dextrose, 1.0 g/L
dipotassium hydrogen phosphate, 3.68 g/L
meat extract, 1.5 g/L
peptone (from meat), 5.0 g/L
potassium dihydrogen phosphate, 1.32 g/L
sodium chloride, 3.5 g/L
yeast extract, 1.5 g/L

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

final pH

7.4±0.2 (25 °C)

application(s)

clinical testing
food and beverages
life science and biopharma
pharmaceutical

microbiology

suitability

bacteria
nonselective for Escherichia coli
nonselective for Klebsiella spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for coliforms

Application

Used in antibiotic assay work (USP, FDA, Ph Eur) according to Grove & Randall; used in the turbidimetric assay of e.g. penicillin, bacitracin and kanamycin with S. aureus and for the cylinder assay of e.g. erythromycin and chloramphenicol with M. inteus.

Preparation Note

Dissolve 17.5 g in 1 litre of warm (60 C) distilled water and sterilize by autoclaving at 121°C for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.C. Grove et al.
Assay Methods of Antibiotics, Med. Encycl. (1955)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service