Skip to Content
Merck
All Photos(1)

Key Documents

A1716

Millipore

Antibiotic assay medium No. 10

suitable for microbiology, NutriSelect® Plus

Sign Into View Organizational & Contract Pricing

Select a Size

500 G
R$2,101.00

R$2,101.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

Select a Size

Change View
500 G
R$2,101.00

About This Item

UNSPSC Code:
41171504
NACRES:
NA.74

R$2,101.00


Estimated to ship onJune 02, 2025


Request a Bulk Order

sterility

non-sterile

Quality Level

product line

BioChemika

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbe id | susceptibility testing: suitable
microbiological culture: suitable

pH

7.2±0.2 (25 °C)

application(s)

clinical testing
food and beverages

microbiology

suitability

bacteria
nonselective for Bordetella spp.
nonselective for Pseudomonas spp.

Application

Used in the performance of antibiotic assays.

Components

Components in g/L:
Casein Enzymic Hydrolysate, 17.00
Papaic Digest of Soyabean Meal, 3.00
Dextrose, 2.50
Sodium Chloride, 5.00
Dipotassium Phosphate, 2.50
Polysorbate 80, 10.00
Agar, 12.00

Preparation Note

Suspend 52 g in 1000 ml distilled water. Heat to dissolve the medium completely. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tests and Methods of Assay of Antibiotics and Antibiotic Containing Drugs, 21, 436-436 (1983)
USP 23/NF18.
United States Pharmacopeia/National Formulary (1995)
USP 22/NF 17.
United States Pharmacopeia/National Formulary (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service