Skip to Content
Merck
All Photos(1)

Key Documents

53798

Supelco

Multi Anion Standard 2 for IC

TraceCERT®, in water

Sign Into View Organizational & Contract Pricing

Select a Size

100 ML
R$1,036.00

R$1,036.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 ML
R$1,036.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$1,036.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

composition

bromide, 1 g/L
chloride, 1 g/L
fluoride, 1 g/L

concentration

in water

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.
Certified content incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com.

Linkage

Visit the IC Portal to learn more

Analysis Note

For the preparation of this CRM only materials of highest purity were used.

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rita de Cássia de Souza et al.
Molecules (Basel, Switzerland), 25(7) (2020-04-08)
Grape seeds are an important byproduct from the grape process. The objective of this work was to evaluate the effect of experimental parameters (temperature and time of pretreatment with ultrasound) to obtain grape seed oil using low pressure (Soxhlet-Sox and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service