Skip to Content
Merck
All Photos(1)

Documents

69523

Supelco

Sodium bicarbonate/Sodium carbonate concentrate

Na2CO3 36 mM and NaHCO3 34 mM in water, IC eluent concentrate (20x) for Metrosep A Supp 4

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NB.21

Quality Level

form

liquid

quality

Na2CO3 36 mM and NaHCO3 34 mM in water

shelf life

limited shelf life, expiry date on the label

concentration

34 mM NaHCO3 in H2O
36 mM Na2CO3 in H2O

technique(s)

ion chromatography: suitable

General description

This eluent concentrate is application tested. It is certified in accordance with ISO Guide 31. All details about exact content, uncertainty, traceability and expiry date are described in the certificate.

Application

Metrohm IC application note AN-S-357: Waste water from a waste water cleaning plant: Anion determination with a Metrosep A Supp 4 - 250/2.0.

Metrohm IC application note AN-U-012: Trace iodide in bottled water using anion chromatography with UV/VIS detection.

Linkage

Visit the IC Portal to learn more

Preparation Note

For the preparation of this CRM only materials of highest purity were used.

Analysis Note

Na2CO3 36 mM + NaHCO3 34 mM in water

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Won Jong Kim et al.
Cell host & microbe, 26(2), 228-239 (2019-08-06)
The mucosa is colonized with commensal Neisseria. Some of these niches are sites of infection for the STD pathogen Neisseria gonorrhoeae (Ngo). Given the antagonistic behavior of commensal bacteria toward their pathogenic relatives, we hypothesized that commensal Neisseria may negatively

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service