Skip to Content
Merck
All Photos(1)

Key Documents

89316

Supelco

Multielement Ion Chromatography Cation Standard Solution, certified

TraceCERT®, suitable for ion chromatography, certified reference material traceable to SI, 10.0 mg/kg Li+, Na+, K+, Mg2+, Ca2+ each cation

Synonym(s):

Certified Multielement Ion Chromatography Cation Standard Solution

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
R$2,527.00

R$2,527.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 ML
R$2,527.00

About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

R$2,527.00


Check Cart for Availability

Request a Bulk Order

grade

certified reference material
TraceCERT®

product line

TraceCERT®

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

technique(s)

ion chromatography: suitable

application(s)

environmental
food and beverages

format

multi-component solution

General description

Certified multiion standard for Ion Chromatography: high purity starting materials

Linkage

Visit the IC Portal to learn more

Analysis Note

Produced with certified high purity materials (Li2CO3, NaCl, KCl, MgO, CaCO3), ultrapure water (18 MOhm, 0.22 mm filtered) and hydrochloric acid

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service