Skip to Content
Merck
All Photos(1)

Documents

30631

Supelco

Sodium 1-decanesulfonate

suitable for ion pair chromatography, LiChropur, ≥99.0%

Synonym(s):

1-Decanesulfonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8CH2SO3Na
CAS Number:
Molecular Weight:
244.33
Beilstein:
3918920
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NB.21

description

anionic

Quality Level

Assay

≥99.0% (T)
≥99.0%

quality

LiChropur

mol wt

244.33 g/mol

technique(s)

ion pair chromatography: suitable

mp

>300 °C (lit.)

λ

5 % in H2O

UV absorption

λ: 210 nm Amax: 0.05
λ: 220 nm Amax: 0.03
λ: 230 nm Amax: 0.02
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

[Na+].CCCCCCCCCCS([O-])(=O)=O

InChI

1S/C10H22O3S.Na/c1-2-3-4-5-6-7-8-9-10-14(11,12)13;/h2-10H2,1H3,(H,11,12,13);/q;+1/p-1

InChI key

AIMUHNZKNFEZSN-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Ion-associating reagent for HPLC, including analyses of peptides and proteins.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Miriam Gochin et al.
Journal of medicinal chemistry, 52(14), 4338-4344 (2009-06-19)
The hydrophobic pocket within the coiled coil domain of HIV-1 gp41 is considered to be a hot-spot suitable for small molecule intervention of fusion, although so far it has yielded only microM inhibitors. Previous peptide studies have identified specific hydrophobic
Luigi Battaglia et al.
Journal of pharmaceutical sciences, 103(7), 2157-2165 (2014-05-16)
The major obstacle to glioblastoma pharmacological therapy is the overcoming of the blood-brain barrier (BBB). In literature, several strategies have been proposed to overcome the BBB: in this experimental work, solid lipid nanoparticles (SLN), prepared according to fatty acid coacervation
Zita Pöstényi et al.
Journal of pharmaceutical and biomedical analysis, 102, 529-534 (2014-12-03)
Validated HPLC analysis was developed in order to monitor the level of 4-fluorodeprenyl in rats. Male Wistar rats were intraperitoneally treated with 30 mg/kg of (-)-4-fluorodeprenyl. The rats were sacrificed after 5, 15, 30 and 60 min of treatment, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service