Skip to Content
Merck
All Photos(1)

Key Documents

51832

Supelco

Sodium 1-heptanesulfonate monohydrate

suitable for ion pair chromatography, LiChropur, ≥99.0% (T)

Synonym(s):

1-Heptanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
R$962.00
50 G
R$3,567.00

R$962.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

Select a Size

Change View
10 G
R$962.00
50 G
R$3,567.00

About This Item

Linear Formula:
CH3(CH2)5CH2SO3Na · H2O
CAS Number:
Molecular Weight:
220.26
Beilstein:
3731762
EC Number:
MDL number:
UNSPSC Code:
12000000
PubChem Substance ID:
NACRES:
NB.21

R$962.00


Available to ship onApril 07, 2025Details


Request a Bulk Order

description

anionic

Quality Level

Assay

≥99.0% (T)

form

solid

quality

LiChropur

mol wt

220.26 g/mol

technique(s)

ion pair chromatography: suitable

λ

10 % in H2O

UV absorption

λ: 210 nm Amax: 0.1
λ: 220 nm Amax: 0.06
λ: 230 nm Amax: 0.04
λ: 260 nm Amax: 0.02
λ: 500 nm Amax: 0.02

suitability

corresponds to standard for RP gradient test
corresponds to standard for filter test

SMILES string

O.[Na+].CCCCCCCS([O-])(=O)=O

InChI

1S/C7H16O3S.Na.H2O/c1-2-3-4-5-6-7-11(8,9)10;;/h2-7H2,1H3,(H,8,9,10);;1H2/q;+1;/p-1

InChI key

XWZCREJRXRKIRQ-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Other Notes

Sodium 1-heptanesulfonate monohydrate has been used as an ion-pairing (IP) reagent, to develop ion-pair reversed phase liquid chromatography method, for the determination of antituberculosis drug ethambutol hydrochloride.[1]

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Runzhong Fu et al.
International journal of molecular sciences, 22(4) (2021-02-11)
Altered lacrimal gland (LG) secretion is a feature of autoimmune dacryoadenitis in Sjögren's syndrome (SS). Cathepsin S (CTSS) is increased in tears of SS patients, which may contribute to disease. Rab3D and Rab27a/b isoforms are effectors of exocytosis in LG
Runzhong Fu et al.
Scientific reports, 10(1), 1455-1455 (2020-01-31)
Autoimmune dacryoadenitis and altered lacrimal gland (LG) secretion are features of Sjögren's syndrome (SS). Activity of cathepsin S (CTSS), a cysteine protease, is significantly and specifically increased in SS patient tears. The soluble chemokine, CX3CL1 (fractalkine), is cleaved from membrane-bound
Marisa Freitas et al.
Phytochemistry, 110, 91-103 (2015-01-03)
The intensification of agricultural productivity is an important challenge worldwide. However, environmental stressors can provide challenges to this intensification. The progressive occurrence of the cyanotoxins cylindrospermopsin (CYN) and microcystin-LR (MC-LR) as a potential consequence of eutrophication and climate change is
Determination of ethambutol by ion-pair reversed phase liquid chromatography with UV detection.
Analytica Chimica Acta, 456.2, 189-192 (2002)

Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service