Skip to Content
Merck
All Photos(1)

Key Documents

860075W

Avanti

C17:0 GD1a (d18:1/17:0)

Avanti Research - A Croda Brand

Synonym(s):

C17:0 GD1a Ganglioside (d18:1/17:0, ammonium salt)

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
R$7,882.00

R$7,882.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

Select a Size

Change View
100 μG
R$7,882.00

About This Item

Empirical Formula (Hill Notation):
C83H149N5O39
CAS Number:
Molecular Weight:
1841.08
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

R$7,882.00


Estimated to ship onApril 19, 2025


Request a Bulk Order

Assay

>99% (TLC)

form

methanol solution

packaging

ampule of 1 × 1 mL

manufacturer/tradename

Avanti Research - A Croda Brand

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C (−15°C to −25°C)

Packaging

2 mL Amber Glass Sealed Ampule (860075W-100UG)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service