Skip to Content
Merck
All Photos(1)

Documents

860683P

Avanti

12:0 Dihydro SM (d18:0/12:0)

Avanti Research - A Croda Brand 860683P, powder

Synonym(s):

N-lauroyl-D-erythro-sphinganylphosphorylcholine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H73N2O6P
CAS Number:
Molecular Weight:
648.94
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 10 mg (860683P-10mg)
pkg of 1 × 5 mg (860683P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 860683P

lipid type

sphingolipids

shipped in

dry ice

storage temp.

−20°C

SMILES string

[H][C@](CCCCCCCCCCCCCCC)(O)[C@@]([H])(NC(CCCCCCCCCCC)=O)COP([O-])(OCC[N+](C)(C)C)=O

InChI

1S/C35H73N2O6P/c1-6-8-10-12-14-16-17-18-19-21-22-24-26-28-34(38)33(32-43-44(40,41)42-31-30-37(3,4)5)36-35(39)29-27-25-23-20-15-13-11-9-7-2/h33-34,38H,6-32H2,1-5H3,(H-,36,39,40,41)/t33-,34+/m0/s1

InChI key

AAKFURSPBCUAIO-SZAHLOSFSA-N

General description

N-lauroyl-D-erythro-sphinganylphosphorylcholine (12:0 Dihydro SM (d18:0/12:0)) is a synthetic sphingomyelin (SM). Dihydro SM contains sphinganine as a long chain base.

Application

N-lauroyl-D-erythro-sphinganylphosphorylcholine (12:0 Dihydro SM (d18:0/12:0)) has been used as an internal standard in electrospray ionization mass spectrometry (ESI-MS) for quantification of lipids extracted from erythrocyte samples.

Packaging

5 mL Amber Glass Screw Cap Vial (860683P-10mg)
5 mL Amber Glass Screw Cap Vial (860683P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Kuikka et al.
Biophysical journal, 80(5), 2327-2337 (2001-04-28)
We have prepared acyl chain-defined D-erythro-sphingomyelins and D-erythro-dihydrosphingomyelins and compared their properties in monolayer and bilayer membranes. Surface pressure/molecular area isotherms of D-erythro-N-16:0-sphingomyelin (16:0-SM) and D-erythro-N-16:0-dihydrosphingomyelin (16:0-DHSM) show very similar packing properties, except that the expanded-to-condensed phase transition (crystallization) occurs
Ayedh Alqarni et al.
Schizophrenia research (2019-10-14)
Omega-3 long chain polyunsaturated fatty acids (n-3 LCPUFA) are necessary for optimum mental health, with recent studies showing low n-3 LCPUFA in people at ultra-high risk (UHR) of developing psychosis. Furthermore, people at UHR of psychosis had increased erythrocyte sphingomyelin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service