Skip to Content
Merck
All Photos(1)

Documents

E34307

Sigma-Aldrich

Ethyl thioglycolate

97%

Synonym(s):

Ethyl mercaptoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2COOC2H5
CAS Number:
Molecular Weight:
120.17
Beilstein:
605581
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.457 (lit.)

bp

54 °C/12 mmHg (lit.)

density

1.096 g/mL at 25 °C (lit.)

SMILES string

CCOC(=O)CS

InChI

1S/C4H8O2S/c1-2-6-4(5)3-7/h7H,2-3H2,1H3

InChI key

PVBRSNZAOAJRKO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

140.9 °F

Flash Point(C)

60.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sha Sha et al.
Journal of agricultural and food chemistry, 65(1), 123-131 (2016-12-19)
The aroma profile of Chinese roasted sesame-like flavor type liquor was investigated by means of headspace solid phase microextraction-aroma extract dilution analysis (HS-SPME-AEDA). Sixty-three odor-active regions were detected by HS-SPME-AEDA with flavor dilution (FD) factors >5, and 58 of these
Timothy M O'Shea et al.
Advanced healthcare materials, 4(12), 1802-1812 (2015-06-20)
Hydrogels with covalently incorporated trehalose are synthesized using thiol-ene Michael addition. Trehalose hydrogels afford prolonged stabilization and -controlled release of model enzymes in vitro and in vivo as well as preservation of protein stability under heat and -lyophilization stressors. Strong

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service