Skip to Content
Merck
All Photos(3)

Key Documents

915297

Sigma-Aldrich

4CzIPN

≥99.9%

Synonym(s):

1,2,3,5-Tetrakis(carbazol-9-yl)-4,6-dicyanobenzene, 2,4,5,6-Tetrakis(9H-carbazol-9-yl) isophthalonitrile

Sign Into View Organizational & Contract Pricing

Select a Size

50 MG
R$2,491.00
250 MG
R$8,151.00

R$2,491.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
50 MG
R$2,491.00
250 MG
R$8,151.00

About This Item

Empirical Formula (Hill Notation):
C56H32N6
CAS Number:
Molecular Weight:
788.89
MDL number:
UNSPSC Code:
12352103
NACRES:
NA.22

R$2,491.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99.9%

form

powder

reaction suitability

reaction type: Photocatalysis
reagent type: catalyst

SMILES string

[n]%11(c%12c(c%13c%11cccc%13)cccc%12)c1c(c(c(c(c1[n]8c9c(c%10c8cccc%10)cccc9)C#N)[n]5c6c(c7c5cccc7)cccc6)C#N)[n]2c3c(c4c2cccc4)cccc3

InChI

1S/C56H32N6/c57-33-43-53(59-45-25-9-1-17-35(45)36-18-2-10-26-46(36)59)44(34-58)55(61-49-29-13-5-21-39(49)40-22-6-14-30-50(40)61)56(62-51-31-15-7-23-41(51)42-24-8-16-32-52(42)62)54(43)60-47-27-11-3-19-37(47)38-20-4-12-28-48(38)60/h1-32H

InChI key

PRWATGACIORDEL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
750050932434908193
4CzIPN ≥99.9%

915297

4CzIPN

TcTa ≥99% (HPLC)

932434

TcTa

3CzClIPN

908193

3CzClIPN

form

powder

form

solid

form

-

form

powder or crystals

reaction suitability

reaction type: Photocatalysis

reaction suitability

-

reaction suitability

-

reaction suitability

reaction type: Photocatalysis, reagent type: catalyst

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

-

Application

4CzIPN is cyanoarene-based donor-acceptor photocatalyst. This highly stable, versatile organic photocatalyst has an excellent redox window allowing for a broad range of applications. It can also be used in the manufacturing of OLEDs.

Product can be used with our line of photoreactors: Including Penn PhD (Z744035) & SynLED 2.0 (Z744080)

Legal Information

Sold in collaboration with Kyulux, Inc.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hiroki Uoyama et al.
Nature, 492(7428), 234-238 (2012-12-14)
The inherent flexibility afforded by molecular design has accelerated the development of a wide variety of organic semiconductors over the past two decades. In particular, great advances have been made in the development of materials for organic light-emitting diodes (OLEDs)
Yong Yao Loh et al.
Science (New York, N.Y.), 358(6367), 1182-1187 (2017-11-11)
Deuterium- and tritium-labeled pharmaceutical compounds are pivotal diagnostic tools in drug discovery research, providing vital information about the biological fate of drugs and drug metabolites. Herein we demonstrate that a photoredox-mediated hydrogen atom transfer protocol can efficiently and selectively install

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service