Skip to Content
Merck
All Photos(1)

Key Documents

669148

Sigma-Aldrich

2-Ethylhexanethiol

97%

Synonym(s):

2-Ethyl-1-hexanethiol, 2-Ethylhexylmercaptan

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
R$428.00

R$428.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
R$428.00

About This Item

Empirical Formula (Hill Notation):
C8H18S
CAS Number:
Molecular Weight:
146.29
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

R$428.00


Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

refractive index

n20/D 1.457

density

0.843 g/mL at 25 °C

SMILES string

CCCCC(CC)CS

InChI

1S/C8H18S/c1-3-5-6-8(4-2)7-9/h8-9H,3-7H2,1-2H3

InChI key

UCJMHYXRQZYNNL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Ethylhexanethiol is an organothiol that forms a self-assembled monolayer (SAM) on a variety of surfaces such as metal oxides and metals. It modifies the surface by acting as a corrosion inhibitor and increases the stability of the surface.[1][2]

Application

2-Ethylhexanethiol can be used as a capping agent on the gold surface for the development of chemiresistive vapor sensors. It also forms a SAM on copper indium sulfide (CuInS2) which can be used as an active layer on organic electronic devices such as organic light emitting diodes (OLEDs) and solar cells.[3][4][5]

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Flam. Liq. 3 - Resp. Sens. 1

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-Throughput Fabrication and Screening Improves Gold Nanoparticle Chemiresistor Sensor Performance
Hubble LJ, et al.
ACS Combinatorial Science, 17(2), 120-129 (2015)
Thermal Stability of Thiolate Self-Assembled Monolayers on Copper Surface
Advanced Materials Research, 646, 18-23 (2013)
Response and discrimination performance of arrays of organothiol-capped Au nanoparticle chemiresistive vapor sensors
Garcia-Berrios E, et al.
The Journal of Physical Chemistry C, 115(14), 6208-6217 (2011)
Communication: Slow supramolecular mode in amine and thiol derivatives of 2-ethyl-1-hexanol revealed by combined dielectric and shear-mechanical studies
J. Chem. Phys. , 646, 18-23 (2015)
High luminance of CuInS 2-based yellow quantum dot light emitting diodes fabricated by all-solution processing
Li J, et al.
Royal Society of Chemistry Advances, 6(76), 72462-72470 (2016)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service