Key Documents
CLS882114
Pyrex® thermometer adapter
joint: ST/NS 14/20
Synonym(s):
lab equipment accessory
Select a Size
1 EA
R$178.00
R$178.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$178.00
About This Item
UNSPSC Code:
41121815
R$178.00
Check Cart for Availability
Joint
joint: ST/NS14/20
manufacturer/tradename
Corning 8821-14
L
60 mm
General description
One end with a ground joint and the other end tooled to accommodate a thermometer.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)