Key Documents
CLS882119
Pyrex® thermometer adapter
joint: ST/NS 19/22
Synonym(s):
lab equipment accessory
Select a Size
1 EA
R$278.00
R$278.00
Check Cart for Availability
Select a Size
Change View
1 EA
R$278.00
About This Item
UNSPSC Code:
41121815
NACRES:
NB.43
R$278.00
Check Cart for Availability
Joint
joint: ST/NS19/22
manufacturer/tradename
Corning 8821-19
L
57 mm
General description
One end with a ground joint and the other end tooled to accommodate a thermometer.
Legal Information
PYREX is a registered trademark of Corning, Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)