Skip to Content
Merck
All Photos(1)

Documents

A3340

Millipore

AC agar

suitable for microbiology, NutriSelect® Plus

Synonym(s):

All Culture Agar

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171605
NACRES:
NA.85

sterility

non-sterile

Quality Level

form

powder

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

NutriSelect® Plus

technique(s)

microbiological culture: suitable

final pH

7.2±0.2 (25 °C)

application(s)

cleaning products
clinical testing
cosmetics
food and beverages
personal care

microbiology

suitability

nonselective for Clostridium spp.
nonselective for Escherichia coli
nonselective for Neisseria spp.
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms

Application

Supports the growth of aerobic, anaerobic and microaerophilic microorganisms

Components

Ingredients (g/L)
Proteose peptone, 20.00
Beef extract, 3.00
Yeast extract, 3.00
Malt extract, 3.00
Dextrose, 5.00
Ascorbic acid, 0.20
Agar, 1.00

Preparation Note

Suspend 35.2 g of AC Agar in 1000 ml of distilled water. Heat to boiling to dissolve the medium completely. Distribute in tubes or bottles to give the desired depth. Sterilize by autoclaving at 15 lbs. pressure (121°C) for 15 minutes. If medium is not used on the same day, it is advisable to drive off dissolved gases by boiling or steaming in the autoclave. Cool without agitation.

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Streptococci- Overview of Detection, Identification, Differentiation and Cultivation Techniques

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service