Skip to Content
Merck
All Photos(2)

Documents

22089

Millipore

Casein peptone Lecithin Polysorbate Broth

suitable for microbiology, NutriSelect® Plus

Synonym(s):

TAT Broth, Tryptic Soy Lecithin Polysorbate Broth, Tryptone Soya Lecithin Polysorbate Broth

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41171607
NACRES:
NA.85

sterility

non-sterile

form

powder

shelf life

limited shelf life, expiry date on the label

composition

casein peptone, 20.0 g/L
soya lecithin, 5.0 g/L

manufacturer/tradename

NutriSelect® Plus

final pH

7.1-7.5 (25 °C)

application(s)

cleaning products
cosmetics
environmental
food and beverages
personal care
pharmaceutical
sterility testing
surface monitoring

microbiology

suitability

nonselective for Bacillus spp.
nonselective for Candida spp.
nonselective for Clostridium spp.
nonselective for Escherichia coli
nonselective for Staphylococcus spp.
nonselective for Streptococcus spp.
nonselective for bacteria (General Media)
nonselective for coliforms
nonselective for molds (General Media)
nonselective for yeasts (General Media)

Application

For the dilution of samples from pharmaceutical, cosmetic and raw material or end-products, for the purpose of enumeration.

Preparation Note

Suspend 25 g in 960 ml distilled water. Heat in a water-bath at 50°C for approx. 30 minutes until completely dissolved. Add 40 ml polysorbate (TWEEN® 20) and sterilize by autoclaving at 121°C for 15 minutes.

Other Notes

Microbial limit tests

Footnote

We offer two media types: the superior granulated GranuCult® and the cost-efficient powdered NutriSelect® culture media, depending on your needs.
The designations basic, plus, or prime are added to indicate the quality control level, from basic quality control to standard QC plus to prime for full regulatory compliance.

Legal Information

GRANUCULT is a registered trademark of Merck KGaA, Darmstadt, Germany
NutriSelect is a registered trademark of Merck KGaA, Darmstadt, Germany
TWEEN is a registered trademark of Croda International PLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

United States Pharmacopeia (1985)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service