Skip to Content
Merck
All Photos(2)

Key Documents

70161

Millipore

Yeast Extract for Biotechnology and Microbiology

for Biotechnology and Microbiology

Synonym(s):

YE, Baker’s yeast extract

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106212
eCl@ss:
42040000
NACRES:
NA.74

Product Name

Yeast Extract, suitable for microbiology

biological source

Saccharomyces cerevisiae

Quality Level

sterility

non-sterile

form

powder

composition

amino-nitrogen, ~5%
total nitrogen (N), ≥11%

ign. residue

≤15%

loss

≤6% loss on drying

pH

7.0±0.2 (2% in H2O)

solubility

H2O: 2%, clear, yellow

application(s)

environmental
food and beverages

microbiology

storage temp.

2-25°C

suitability

molds
yeasts

Looking for similar products? Visit Product Comparison Guide

General description

A water-soluble extract of autolyzed yeast cells, yeast extract is a mixture of amino acids, peptides, and water-soluble vitamins, for example, intact vitamin B12 complex and carbohydrates, and can be used as an additive for culture media and fermentation media. It can also be used in the resuscitation of a large variety of fastidious microorganisms.

Application

Yeast extract is recommended for use in culture media and the cultivation of various microorganisms. It provides multi-functional nutrient supplements that can also be used in microbial fermentation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Paul Vigne et al.
BMC physiology, 10, 8-8 (2010-05-15)
Recent attention has been given to the relationships between diet, longevity, aging and resistance to various forms of stress. Flies do not simply ingest calories. They sense different concentrations of carbohydrate and protein macronutrients and they modify their feeding behavior
Alfredo Perales-Puchalt et al.
Journal of leukocyte biology, 103(5), 799-805 (2018-03-15)
Due to their cytotoxic activities, many anticancer drugs cause extensive damage to the intestinal mucosa and have antibiotic activities. Here, we show that cisplatin induces significant changes in the repertoire of intestinal commensal bacteria that exacerbate mucosal damage. Restoration of
Emilio M Ungerfeld et al.
Microorganisms, 8(6) (2020-05-30)
Ameliorating methane (CH4) emissions from ruminants would have environmental benefits, but it is necessary to redirect metabolic hydrogen ([H]) toward useful sinks to also benefit animal productivity. We hypothesized that inhibiting rumen methanogenesis would increase de novo synthesis of microbial
Myounghoon Moon et al.
Scientific reports, 9(1), 19959-19959 (2019-12-29)
Sugar refinery washing water (SRWW) contains abundant levels of carbon sources and lower levels of contaminants than other types of wastewater, which makes it ideal for heterotrophic cultivation of microalgae. Here, carbon sources in SRWW were utilized for conversion into
Sara Hooshangi et al.
PLoS computational biology, 7(9), e1002172-e1002172 (2011-10-08)
Quorum sensing (QS) enables bacterial multicellularity and selective advantage for communicating populations. While genetic "switching" phenomena are a common feature, their mechanistic underpinnings have remained elusive. The interplay between circuit components and their regulation are intertwined and embedded. Observable phenotypes

Articles

Culture media provides a habitat with suitable nutrients, energy sources, and certain environmental conditions for the growth of microorganisms. The components of the culture media range from simple sugars to peptones, salts, antibiotics, and complex indicators.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service