Skip to Content
Merck
All Photos(1)

Key Documents

260347

Sigma-Aldrich

1,2-Epoxy-5-hexene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O
CAS Number:
Molecular Weight:
98.14
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

liquid

impurities

<3% benzene

refractive index

n20/D 1.425 (lit.)

bp

119-121 °C (lit.)

density

0.87 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCCC1CO1

InChI

1S/C6H10O/c1-2-3-4-6-5-7-6/h2,6H,1,3-5H2

InChI key

MUUOUUYKIVSIAR-UHFFFAOYSA-N

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Carc. 1A - Flam. Liq. 2 - Muta. 1B - STOT RE 2

Target Organs

Blood

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takuya Miyazaki et al.
Advanced healthcare materials, 9(16), e2000538-e2000538 (2020-06-26)
Messenger RNA (mRNA) shows high therapeutic potential, though effective delivery systems are still needed for boosting its application. Nanocarriers loading mRNA via polyion complexation with block catiomers into core-shell micellar structures are promising systems for enhancing mRNA delivery. Engineering the
Yuanchen Liu et al.
Analytical and bioanalytical chemistry, 410(2), 509-519 (2017-12-01)
A series of β-cyclodextrin derivatives were designed and synthesized. The derivatives were investigated as functional monomers in molecularly imprinted polymeric solid-phase microextraction (MIP-SPME) fiber coatings. The coatings, with a layer thickness of 250 μm, were immobilized onto stainless steel using

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service