Skip to Content
Merck
All Photos(1)

Documents

128554

Sigma-Aldrich

1,5-Hexadiene

97%

Synonym(s):

Biallyl, Diallyl

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCH2CH2CH=CH2
CAS Number:
Molecular Weight:
82.14
Beilstein:
1731106
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

367 mmHg ( 37.7 °C)

Quality Level

Assay

97%

form

liquid

contains

Contains ~100ppm BHT as stabilizer

refractive index

n20/D 1.404 (lit.)

bp

60 °C (lit.)

mp

−141 °C (lit.)

density

0.692 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CCCC=C

InChI

1S/C6H10/c1-3-5-6-4-2/h3-4H,1-2,5-6H2

InChI key

PYGSKMBEVAICCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,5-Hexadiene was used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Björn Nelson et al.
Organic letters, 13(16), 4438-4441 (2011-07-29)
Scope and limitations of the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes have been studied. In situ NMR experiments indicate a challenging competition between various reaction pathways. A careful balance between substrate structure, nature of the precatalyst, and reaction conditions was required
Amber L Russell et al.
International journal of mass spectrometry, 287(1-3), 8-15 (2010-04-20)
A method for the study of reactions of open-shell neutrals (radicals) and radical cations is described. Pyrolysis (25-1500 degrees C) of thermally labile compounds, such as, 1,5-hexadiene via a Chen nozzle yields a seeded beam of reactive species in helium.
Aman Ullah et al.
ChemSusChem, 10(10), 2167-2174 (2017-03-02)
Catalytic transformation of renewable feedstocks into fine chemicals is in high demands and olefin metathesis is a sophisticated tool for biomass conversion. Nevertheless, the large-scale viability of such processes depends on the conversion efficiency, energy efficiency, catalytic activity, selective conversion
Richard G Closser et al.
ACS applied materials & interfaces, 11(24), 21988-21997 (2019-06-11)
The synthesis of an all-carbon backboned fluoropolymer using photoactivated molecular layer deposition (pMLD) is developed. pMLD is a vapor-phase, layer-by-layer, organic thin film synthesis method utilizing UV light, allowing for the creation of materials previously unavailable via thermal MLD. The
He Nan et al.
Journal of chromatography. A, 1523, 316-320 (2017-06-26)
Silver ion or argentation chromatography utilizes stationary phases containing silver ions for the separation of unsaturated compounds. In this study, a mixed-ligand silver-based ionic liquid (IL) was evaluated for the first time as a gas chromatographic (GC) stationary phase for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service