HPLC Analysis of 4-Benzyl-2-oxazolidinone Enantiomers on Astec® CHIROBIOTIC® TAG
CONDITIONS
column
CHIROBIOTIC TAG, 25 cm x 4.6 mm I.D., 5 μm particles (14024AST)
mobile phase
methanol
flow rate
1 mL/min
column temp.
23 °C
detector
UV, 230 nm
injection
2 μL
sample
2 mg/mL in methanol