Merck
Chips de pomme de terre
Chips de pomme de terre suitable for GC, application for SPME
Matériaux
Colonne analytique
Réf. du produit
Description
Tarif
Colonne capillaire SPB®-1 SOUFRE pour GC/CPG
L × I.D. 30 m × 0.32 mm, df 4.00 μm
Fibre SPME
Réf. du produit
Description
Tarif
Ensemble de fibres de SPME, divinylbenzène/carboxène/polydiméthylsiloxane (DVB/CAR/PDMS)
50/30µm DVB/CAR/PDMS, StableFlex (1cm), needle size 24 ga, Manual Holder, pk of 3, plain gray hub
Produit(s) apparenté(s)
Réf. du produit
Description
Tarif
Support de fibres SPME
for use with Manual Sampling, pk of 1
Support de fibres SPME
for use with CTC CombiPAL, Gerstel MPS2 Autosamplers, pk of 1
Étalon
Réf. du produit
Description
Tarif
Decanal
≥98% (GC), liquid
Methyl butyrate
analytical standard
Hexanoic acid
analytical standard
Octanoic acid
analytical standard
Octane
analytical standard
Heptanal
≥95%, FCC, FG
Nonanal
analytical standard
Hexanal
analytical standard
Butyl hexanoate
analytical standard
Octanal
analytical standard
Heptanoic acid
analytical standard
CONDITIONS
sample/matrix
3 g crushed potato chips in 15 mL vial
SPME fiber
DVB/Carboxen/PDMS, 50/30 μm, StableFlex (57328-U)
extraction
headspace, 65 °C, 20 min, with stirring
desorption process
3 min, 250 °C
column
SPB-1 SULFUR, 30 m × 0.32 mm I.D., 4.0 μm (24158)
inj. temp.
250 °C
oven
45 °C (1.5 min) to 250 °C at 12 °C/min, hold 10 min
detector
quadrupole mass spectrometer, m/z = 35-290 at 0.6 sec/scan
carrier gas
helium, 40 cm/sec
injection
Splitless/split, closed 2 min
liner
0.75 mm liner
sample
3g crushed potato chips in 15 mL vial
Description
Remarque sur l'analyse
Potato chips can contain up to 50% oil and are subject to rancidity. Aldehydes, particularly hexanal and heptanal, are formed from fatty acids that are cleaved at the double bonds when exposed to ultraviolet light and air. A package of 3-year-old potato chips was analyzed using SPME and compared to a fresh package. The presence of aldehydes indicated that the older chips were rancid. However, due to the age of the chips, further oxidation of the aldehydes occurred that resulted in the formation of hexanoic and heptanoic acids. A Carboxen/PDMS/DVB fiber was inserted into the headspace of the potato chip samples at 65 °C for 20 minutes with agitation. The fiber was then analyzed using GC/MS on a SPB-1 SULFUR column. This non-polar column has a thick film and is ideal for smaller molecules.
Informations légales
SPB is a registered trademark of Merck KGaA, Darmstadt, Germany