Principaux documents
57364-U
Supelco
Ensemble support d'échantillonnage et tige SPME
greener alternative
for use with SPME Sampling Stand
1 EA
185,00 €
185,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Changer de vue
1 EA
185,00 €
About This Item
Code UNSPSC :
41115854
Nomenclature NACRES :
NB.21
185,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Devis pour commande en gros
Caractéristiques du produit alternatif plus écologique
Waste Prevention
Safer Solvents and Auxiliaries
Learn more about the Principles of Green Chemistry.
sustainability
Greener Alternative Product
Technique(s)
solid phase microextraction (SPME): suitable
Application(s)
food and beverages
Compatibilité
for use with SPME Sampling Stand
Autre catégorie plus écologique
Description générale
We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Greener Chemistry. This is a Solid Phase Microextraction (SPME) technology product. SPME is one of the greener technologies for sample preparation as it is solvent free extraction technology and SPME fibres are re-usable and generates as little waste as possible, thus aligns with "Waste Prevention”, and ″Safer Solvents and Auxiliaries″. Click here for more information.