Principaux documents
33284
Décapsuleur de fiole, type pince
for use with 8mm crimp seals, pkg of 1 ea
About This Item
Code UNSPSC :
41122001
Description
Decapper, pliers style
Conditionnement
pkg of 1 ea
Compatibilité
for use with 8mm crimp seals
Description générale
Decapper provides quick and easy removal of seals.