Principaux documents
Z328774
BRAND® magnetic stirring bar, PTFE, cylindrical
L 25 mm, bar diam. 6 mm
Synonyme(s) :
PTFE Magnetic Stirrer
10 EA
41,00 €
41,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Changer de vue
10 EA
41,00 €
About This Item
Code UNSPSC :
41103806
eCl@ss :
32100100
Nomenclature NACRES :
NB.72
41,00 €
Veuillez contacter notre Service Clients pour connaître la disponibilité de ce produit.
Matériaux
PTFE-coated
cylindrical
Fabricant/nom de marque
BRAND 137120
Longueur
25 mm
Diam. de la barre
6 mm
Description générale
BRAND® magnetic cylindrical stirring bars are expertly designed for optimal mixing performance, featuring a smooth, round shape that enhances stirring efficiency. Their Alnico V magnetic cores are protected by a robust PTFE (polytetrafluoroethylene) coating, ensuring durability and resistance to chemical damage. These stirring bars are particularly effective for use in large volumes or thick mixtures, making them ideal for a variety of laboratory applications.
Caractéristiques et avantages
  • BRAND® magnetic cylindrical stirring bars are round with a smooth surface.
  • The Alnico V magnetic cores are encapsulated with high-grade, crack-resistant PTFE (polytetrafluoroethylene) coating.
  • Ideal for large volumes or highly viscous mixtures.
Autres remarques
BR137120 (India)
Informations légales
BRAND is a registered trademark of BRAND GMBH + CO KG